Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316676.1 | NP_001303605.1 | 22 | 3980 | ||
NM_001323354.1 | NP_001310283.1 | 20 | 5373 | |||
NM_001323355.1 | NP_001310284.1 | 20 | 3786 | |||
NM_001323356.1 | NP_001310285.1 | 20 | 3780 | |||
NM_001323357.1 | NP_001310286.1 | 17 | 3365 | |||
NM_006504.5 | NP_006495.1 | 21 | 3809 | |||
NM_130435.4 | NP_569119.1 | 18 | 3456 | |||
XM_011539996.1 | XP_011538298.1 | 19 | 3645 | |||
GenBank mRNA | AK001335.1 | - | 2 | 1563 | ||
AK026334.1 | - | 1 | 369 | |||
DA949010.1 | - | 1 | 551 | |||
H17674.1 | - | 1 | 192 |