Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006741.3 | NP_006732.3 | 6 | 630 | ||
XM_005268995.4 | XP_005269052.1 | 5 | 454 | |||
XM_006719471.3 | XP_006719534.1 | 4 | 404 | |||
GenBank mRNA | AK123969.1 | - | 5 | 335 | ||
AY063766.1 | - | 6 | 461 | |||
BC022470.1 | - | 6 | 630 | 4819907 | ||
CR457003.1 | - | 6 | 459 | |||
U48707.1 | - | 6 | 467 |