Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006813.2 | NP_006804.1 | 2 | 1499 | ||
GenBank mRNA | AF279899.1 | - | 1 | 494 | ||
AK130046.1 | - | 2 | 1457 | |||
AK130722.1 | - | 1 | 1539 | |||
BC018112.1 | - | 2 | 1369 | 3916954 | ||
BC044919.1 | - | 2 | 1679 | |||
U03105.1 | - | 2 | 1427 |