Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006951.4 | NP_008882.2 | 2 | 655 | ||
NM_139052.2 | NP_620640.1 | 2 | 655 | |||
GenBank mRNA | AK223615.1 | - | 2 | 627 | ||
AK291229.1 | - | 2 | 655 | |||
BC052268.1 | - | 2 | 633 | 6575596 | ||
BC136340.1 | - | 2 | 636 | 9020327 | ||
BC136348.1 | - | 2 | 636 | 9020335 | ||
U75309.1 | - | 2 | 621 | |||
U80191.1 | - | 2 | 632 | |||
X95525.1 | - | 2 | 641 |