Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318933.1 | NP_001305862.1 | 24 | 2111 | ||
NM_002390.5 | NP_002381.2 | 24 | 2115 | |||
XM_005257373.3 | XP_005257430.1 | 24 | 2257 | |||
XM_017024647.1 | XP_016880136.1 | 23 | 2029 | |||
GenBank mRNA | AB009675.1 | - | 24 | 2094 | ||
AK296512.1 | - | 24 | 2192 | |||
D17390.1 | - | 21 | 1834 |