Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU6F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166018.1 | NP_001159490.1 | 7 | 1089 | ||
NM_007252.3 | NP_009183.3 | 7 | 1089 | |||
XM_011515113.2 | XP_011513415.1 | 6 | 1109 | |||
GenBank mRNA | U91935.1 | - | 6 | 1032 |