Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206774.1 | NP_001193703.1 | 5 | 867 | ||
NM_012086.4 | NP_036218.1 | 5 | 867 | |||
XM_005246965.4 | XP_005247022.1 | 5 | 782 | |||
GenBank mRNA | AF133123.1 | - | 5 | 803 | ||
AF465407.1 | - | 5 | 758 | |||
AK025560.1 | - | 5 | 781 | |||
AK314718.1 | - | 5 | 808 | |||
AL137453.1 | - | 2 | 209 | |||
BC043347.1 | - | 5 | 722 | 5498609 | ||
S81493.1 | - | 2 | 229 |