Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012476.2 | NP_036608.1 | 2 | 349 | ||
XM_006711982.3 | XP_006712045.1 | 2 | 388 | |||
XM_011532750.2 | XP_011531052.1 | 2 | 388 | |||
XM_011532751.2 | XP_011531053.1 | 2 | 388 | |||
GenBank mRNA | AB593145.1 | - | 2 | 358 | ||
BC006336.2 | - | 2 | 324 | 4304066 | ||
BT007035.1 | - | 2 | 317 | |||
Y17791.1 | - | 2 | 349 |