Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282470.2 | NP_001269399.1 | 18, 19 | 2743 | ||
NM_014619.4 | NP_055434.2 | 19, 20 | 2703 | |||
XM_011542784.1 | XP_011541086.1 | 17, 18 | 2399 | |||
XM_011542786.2 | XP_011541088.1 | 16, 17 | 3203 | |||
XM_011542787.2 | XP_011541089.1 | 12, 13 | 3653 | |||
XM_017017621.1 | XP_016873110.1 | 17, 18 | 2387 | |||
XM_017017622.1 | XP_016873111.1 | 17, 18 | 2357 | |||
GenBank mRNA | AK292726.1 | - | 19, 20 | 2690 | ||
AK314284.1 | - | 19, 20 | 2745 | |||
S67803.1 | - | 17, 18 | 2403 |