Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014621.2 | NP_055436.2 | 2 | 1010 | ||
XM_005246514.4 | XP_005246571.1 | 2 | 1904 | |||
GenBank mRNA | AK311735.1 | - | 2 | 440 | ||
BC016763.1 | - | 2 | 828 | 4068219 | ||
BC074797.2 | - | 2 | 808 | 30915295 | ||
X04706.1 | - | 2 | 1010 |