Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136264.1 | NP_001129736.1 | 5 | 878 | ||
NM_001136266.1 | NP_001129738.1 | 4 | 867 | |||
NM_001136267.1 | NP_001129739.1 | 4 | 848 | |||
NM_001136268.1 | NP_001129740.1 | 3 | 698 | |||
NM_014764.3 | NP_055579.1 | 4 | 944 | |||
GenBank mRNA | AK095963.1 | - | 5 | 737 | ||
AK125855.1 | - | 2 | 2591 | |||
AK290119.1 | - | 4 | 844 | |||
AK303828.1 | - | 4 | 767 | |||
AY430097.1 | - | 4 | 843 | |||
BC002334.2 | - | 4 | 826 | 3509229 | ||
BC007900.2 | - | 4 | 825 | 4297784 | ||
BX647278.1 | - | 4 | 842 | |||
D31767.1 | - | 4 | 829 |