Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304482.1 | NP_001291411.1 | 13, 14 | 1180 | ||
NM_015466.3 | NP_056281.1 | 14, 15 | 1308 | |||
XM_005265031.2 | XP_005265088.2 | 14, 15 | 1259 | |||
GenBank mRNA | AB025194.1 | - | 14, 15 | 1256 | ||
AB040904.2 | - | 14, 15 | 1308 | |||
AF290614.1 | - | 14, 15 | 1254 | |||
AK055157.1 | - | 14, 15 | 1279 | |||
AK289502.1 | - | 14, 15 | 1257 | |||
AK295877.1 | - | 14, 15 | 1228 | |||
AK295888.1 | - | 13, 14 | 1171 | |||
AK299906.1 | - | 13, 14 | 1151 | |||
BC027711.2 | - | 3, 4 | 247 | |||
BC089042.1 | - | 14, 15 | 1226 | 6579163 |