Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005387.6 | NP_005378.4 | 3, 4 | 604 | ||
NM_016320.4 | NP_057404.2 | 3, 4 | 604 | |||
NM_139131.4 | NP_624357.1 | 3, 4 | 604 | |||
NM_139132.3 | NP_624358.2 | 3, 4 | 604 | |||
GenBank mRNA | AB040538.1 | - | 3, 4 | 359 | ||
AF071076.1 | - | 3, 4 | 307 | |||
AF071077.1 | - | 3, 4 | 307 | |||
AF231130.1 | - | 3, 4 | 327 | |||
AK292882.1 | - | 3, 4 | 344 | |||
AK301512.1 | - | 3, 4 | 364 | |||
BC041136.1 | - | 3, 4 | 353 | 5179730 | ||
KU847959.1 | - | 183 | ||||
U41815.1 | - | 3, 4 | 327 |