Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016322.3 | NP_057406.2 | 4 | 677 | ||
GenBank mRNA | AF112206.1 | - | 4 | 285 | ||
AF152463.1 | - | 3 | 221 | |||
AF203689.1 | - | 3 | 227 | |||
AF498949.1 | - | 3 | 221 | |||
AK023524.1 | - | 4 | 459 | |||
AK090889.1 | - | 4 | 476 | |||
AL162081.1 | - | 4 | 404 | |||
AY553875.1 | - | 3 | 221 | |||
BC006081.1 | - | 4 | 448 | 2963119 | ||
CR457394.1 | - | 3 | 221 |