Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323625.1 | NP_001310554.1 | 9 | 1513 | ||
NM_016361.4 | NP_057445.4 | 9 | 1482 | |||
NR_136633.1 | 9 | 1468 | ||||
NR_136634.1 | 10 | 1843 | ||||
NR_136635.1 | 9 | 1468 | ||||
NR_136636.1 | 9 | 1788 | ||||
XM_011509601.2 | XP_011507903.1 | 9 | 1438 | |||
XM_017001422.1 | XP_016856911.1 | 9 | 1438 | |||
GenBank mRNA | AB030039.2 | - | 9 | 1286 | ||
AB031478.1 | - | 9 | 1393 | |||
AB209248.1 | - | 8 | 3564 | |||
AK000657.1 | - | 9 | 1219 | |||
AY358552.1 | - | 9 | 1190 | |||
BC009965.1 | - | 9 | 1462 | 4299943 | ||
BC034686.1 | - | 9 | 1466 | 3463770 | ||
CR457101.1 | - | 9 | 997 | |||
KJ534759.1 | - | 9 | 1018 | |||
KJ535036.1 | - | 8 | 890 | |||
KJ535044.1 | - | 6 | 656 | |||
KJ535055.1 | - | 9 | 973 |