Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017409.3 | NP_059105.2 | 1 | 413 | ||
GenBank mRNA | AF255675.1 | - | 1 | 315 | ||
AK313759.1 | - | 1 | 429 | |||
AL558161.3 | - | 1 | 413 | |||
BC001293.1 | - | 1 | 386 | 3458115 | ||
BT009921.1 | - | 1 | 315 |