Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017820.4 | NP_060290.3 | 19 | 2340 | ||
XM_011518810.1 | XP_011517112.1 | 21 | 2715 | |||
XM_011518814.1 | XP_011517116.1 | 13 | 1665 | |||
XM_011518815.1 | XP_011517117.1 | 10 | 1349 | |||
XM_017014861.1 | XP_016870350.1 | 22 | 2859 | |||
XM_017014862.1 | XP_016870351.1 | 20 | 2600 | |||
XM_017014863.1 | XP_016870352.1 | 20 | 2558 | |||
XM_017014864.1 | XP_016870353.1 | 19 | 2920 | |||
XM_017014865.1 | XP_016870354.1 | 19 | 2567 | |||
XM_017014866.1 | XP_016870355.1 | 18 | 2223 | |||
XM_017014867.1 | XP_016870356.1 | 18 | 2264 | |||
GenBank mRNA | AB061798.1 | - | 13 | 1689 | ||
AK055004.1 | - | 9 | 1472 | |||
AK094438.1 | - | 19 | 2240 | |||
BU947354.1 | - | 1 | 62 |