Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002410.4 | NP_002401.1 | 1 | 330 | ||
XM_005263668.4 | XP_005263725.1 | 2 | 695 | |||
XM_005263669.4 | XP_005263726.1 | 2 | 692 | |||
XM_005263670.3 | XP_005263727.1 | 2 | 348 | |||
XM_006712534.3 | XP_006712597.1 | 3 | 544 | |||
XM_011511199.2 | XP_011509501.1 | 3 | 979 | |||
XM_011511200.1 | XP_011509502.1 | 2 | 344 | |||
XM_011511201.2 | XP_011509503.1 | 2 | 448 | |||
XM_011511202.1 | XP_011509504.1 | 2 | 345 | |||
XM_011511204.2 | XP_011509506.1 | 3 | 547 | |||
XM_017004147.1 | XP_016859636.1 | 3 | 819 | |||
XM_017004148.1 | XP_016859637.1 | 2 | 656 | |||
XM_017004149.1 | XP_016859638.1 | 2 | 697 | |||
GenBank mRNA | AF113921.1 | - | 1 | 218 | ||
D17716.1 | - | 1 | 330 |