Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303034.1 | NP_001289963.1 | 2002 | |||
NM_001303035.1 | NP_001289964.1 | 1935 | ||||
NM_020402.3 | NP_065135.2 | 1940 | ||||
GenBank mRNA | AF199235.2 | - | 1940 | |||
AJ278118.1 | - | 1910 |