Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021129.3 | NP_066952.1 | 11 | 1249 | ||
GenBank mRNA | AB026723.1 | - | 11 | 1172 | ||
AF092439.1 | - | 11 | 1219 | |||
AF119665.1 | - | 11 | 1219 | |||
AF154065.1 | - | 11 | 1187 | |||
AF217186.1 | - | 11 | 1142 | |||
AK058193.1 | - | 11 | 1242 | |||
BC001022.2 | - | 11 | 1224 | 3345496 | ||
BC061581.1 | - | 11 | 1232 | 6526628 | ||
BC107882.1 | - | 11 | 1183 | 6495107 | ||
EU794626.1 | - | 11 | 1249 |