Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021570.3 | NP_067545.3 | 2, 3 | 694 | ||
GenBank mRNA | AF213356.1 | - | 2, 3 | 686 | ||
BC009458.1 | - | 1, 2 | 541 | |||
BC064363.1 | - | 2, 3 | 739 | 5555949 |