Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166247.1 | NP_001159719.1 | 3 | 771 | ||
NM_021956.4 | NP_068775.1 | 3 | 771 | |||
NM_175768.3 | NP_786944.1 | 3 | 771 | |||
XM_005266945.2 | XP_005267002.1 | 4 | 901 | |||
XM_005266946.3 | XP_005267003.1 | 3 | 856 | |||
XM_011535777.2 | XP_011534079.1 | 3 | 938 | |||
XM_011535778.2 | XP_011534080.1 | 3 | 938 | |||
XM_017010781.1 | XP_016866270.1 | 3 | 938 | |||
XM_017010782.1 | XP_016866271.1 | 3 | 938 | |||
GenBank mRNA | AJ252246.1 | - | 3 | 478 | ||
AJ301610.1 | - | 3 | 478 | |||
AK289542.1 | - | 3 | 705 | |||
AK291965.1 | - | 3 | 727 | |||
BC037954.1 | - | 3 | 963 | |||
BC063814.1 | - | 3 | 681 | 6189201 | ||
HM149335.1 | - | 3 | 478 | |||
HM149336.1 | - | 3 | 478 | |||
HM149337.1 | - | 3 | 478 | |||
HM149338.1 | - | 3 | 478 | |||
HM149339.1 | - | 3 | 478 | |||
U16126.1 | - | 3 | 478 |