Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022062.2 | NP_071345.2 | 5, 6 | 535 | ||
XM_005271642.2 | XP_005271699.1 | 4, 5 | 424 | |||
XM_005271643.2 | XP_005271700.1 | 2, 3 | 241 | |||
XM_011542944.2 | XP_011541246.1 | 5, 6 | 620 | |||
XM_011542945.2 | XP_011541247.1 | 6, 7 | 580 | |||
XM_011542946.1 | XP_011541248.1 | 5, 6 | 503 | |||
XM_017018110.1 | XP_016873599.1 | 5, 6 | 3370 | |||
XM_017018112.1 | XP_016873601.1 | 2, 3 | 241 | |||
GenBank mRNA | AB065001.1 | - | 2, 3 | 213 | ||
AJ417081.1 | - | 2, 3 | 185 | |||
AK023136.1 | - | 2, 3 | 213 | |||
AK295857.1 | - | 4, 5 | 402 | |||
AK316001.1 | - | 4, 5 | 335 | |||
AK316268.1 | - | 3, 4 | 323 | |||
BC045626.1 | - | 5, 6 | 492 | 5261106 | ||
BX648615.1 | - | 3, 4 | 312 | |||
DA789677.1 | - | 3, 4 | 357 |