Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022076.3 | NP_071359.3 | 1 | 318 | ||
GenBank mRNA | AF533018.1 | - | 1 | 320 | ||
AY156515.1 | - | 1 | 317 | |||
BC119755.1 | - | 1 | 266 | 40119299 | ||
BC119756.1 | - | 1 | 266 | 40119300 | ||
CR457159.1 | - | 1 | 188 |