Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290360.2 | NP_001277289.1 | 3 | 603 | ||
NM_001324101.1 | NP_001311030.1 | 3 | 603 | |||
NM_001324103.1 | NP_001311032.1 | 3 | 603 | |||
NM_001324106.1 | NP_001311035.1 | 3 | 603 | |||
NM_001324107.1 | NP_001311036.1 | 3 | 603 | |||
NM_001324108.1 | NP_001311037.1 | 3 | 603 | |||
NM_001324109.1 | NP_001311038.1 | 3 | 603 | |||
NM_001324111.1 | NP_001311040.1 | 3 | 603 | |||
NM_024007.4 | NP_076870.1 | 3 | 603 | |||
NM_182708.2 | NP_874367.1 | 3 | 603 | |||
XM_017009192.1 | XP_016864681.1 | 3 | 603 | |||
XM_017009193.1 | XP_016864682.1 | 3 | 591 | |||
XM_017009194.1 | XP_016864683.1 | 3 | 603 | |||
XM_017009195.1 | XP_016864684.1 | 3 | 603 | |||
XM_017009196.1 | XP_016864685.1 | 3 | 1538 | |||
XM_017009197.1 | XP_016864686.1 | 3 | 1044 | |||
XM_017009198.1 | XP_016864687.1 | 3 | 963 | |||
XM_017009202.1 | XP_016864691.1 | 3 | 603 | |||
XM_017009203.1 | XP_016864692.1 | 3 | 603 | |||
GenBank mRNA | AF208502.1 | - | 3 | 305 | ||
AK289712.1 | - | 3 | 590 | |||
AK303698.1 | - | 1 | 229 | |||
AK304438.1 | - | 3 | 306 | |||
BC038805.1 | - | 3 | 614 | 4524661 | ||
BC041178.1 | - | 3 | 530 | 6045572 |