Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290360.2 | NP_001277289.1 | 3, 4 | 642 | ||
NM_001324101.1 | NP_001311030.1 | 3, 4 | 642 | |||
NM_001324103.1 | NP_001311032.1 | 3, 4 | 642 | |||
NM_001324106.1 | NP_001311035.1 | 3, 4 | 642 | |||
NM_001324107.1 | NP_001311036.1 | 3, 4 | 642 | |||
NM_001324108.1 | NP_001311037.1 | 3, 4 | 642 | |||
NM_001324109.1 | NP_001311038.1 | 3, 4 | 642 | |||
NM_024007.4 | NP_076870.1 | 3, 4 | 642 | |||
NM_182708.2 | NP_874367.1 | 3, 4 | 642 | |||
XM_017009192.1 | XP_016864681.1 | 3, 4 | 642 | |||
XM_017009193.1 | XP_016864682.1 | 3, 4 | 630 | |||
XM_017009194.1 | XP_016864683.1 | 3, 4 | 642 | |||
XM_017009195.1 | XP_016864684.1 | 3, 4 | 642 | |||
XM_017009196.1 | XP_016864685.1 | 3, 4 | 1577 | |||
XM_017009197.1 | XP_016864686.1 | 3, 4 | 1083 | |||
XM_017009198.1 | XP_016864687.1 | 3, 4 | 1002 | |||
XM_017009202.1 | XP_016864691.1 | 3, 4 | 642 | |||
XM_017009203.1 | XP_016864692.1 | 3, 4 | 642 | |||
GenBank mRNA | AF208502.1 | - | 3, 4 | 344 | ||
AK289712.1 | - | 3, 4 | 629 | |||
AK303698.1 | - | 1, 2 | 268 | |||
AK304438.1 | - | 3, 4 | 345 | |||
BC038805.1 | - | 3, 4 | 653 | 4524661 | ||
BC041178.1 | - | 3, 4 | 569 | 6045572 |