Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030791.3 | NP_110418.1 | 3 | 1672 | ||
XM_017021678.1 | XP_016877167.1 | 4 | 1736 | |||
GenBank mRNA | AF349315.1 | - | 3 | 1594 | ||
AJ293294.1 | - | 3 | 1553 | |||
BC063839.1 | - | 3 | 1615 | 6184825 | ||
BC070060.1 | - | 3 | 1632 | 30347615 |