Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002427.3 | NP_002418.1 | 2 | 208 | ||
GenBank mRNA | AK222748.1 | - | 2 | 208 | ||
AK292211.1 | - | 2 | 208 | |||
AK315341.1 | - | 2 | 206 | |||
AY256443.1 | - | 2 | 179 | |||
AY256444.1 | - | 2 | 179 | |||
BC067522.1 | - | 2 | 187 | 7002184 | ||
BC067523.1 | - | 2 | 187 | 7002185 | ||
BC074807.2 | - | 2 | 198 | 30915315 | ||
BC074808.2 | - | 2 | 198 | 30915529 | ||
X75308.1 | - | 2 | 183 | |||
X81334.1 | - | 2 | 206 |