Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007079.3 | NP_009010.2 | 4 | 1758 | ||
NM_032611.2 | NP_116000.1 | 5 | 1833 | |||
XM_005250764.2 | XP_005250821.1 | 7 | 2218 | |||
XM_006716500.2 | XP_006716563.1 | 6 | 1853 | |||
XM_011516803.1 | XP_011515105.1 | 6 | 2095 | |||
XM_011516805.1 | XP_011515107.1 | 7 | 1352 | |||
XM_017012998.1 | XP_016868487.1 | 5 | 1850 | |||
XM_017012999.1 | XP_016868488.1 | 5 | 1778 | |||
GenBank mRNA | AJ276554.1 | - | 3 | 945 | ||
BC003105.1 | - | 4 | 1239 | 3357244 |