Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039970.1 | NP_001035059.1 | 15 | 3025 | ||
NM_001278236.1 | NP_001265165.1 | 15 | 3859 | |||
NM_001278238.1 | NP_001265167.1 | 14 | 2988 | |||
NM_001278239.1 | NP_001265168.1 | 14 | 2892 | |||
NM_006906.1 | NP_008837.1 | 15 | 3121 | |||
NM_032781.3 | NP_116170.3 | 15 | 3076 | |||
XM_011520411.2 | XP_011518713.1 | 15 | 3107 | |||
XM_017018434.1 | XP_016873923.1 | 14 | 2899 | |||
XM_017018435.1 | XP_016873924.1 | 14 | 2869 | |||
XM_017018436.1 | XP_016873925.1 | 13 | 2805 | |||
XM_017018437.1 | XP_016873926.1 | 14 | 2823 | |||
XM_017018438.1 | XP_016873927.1 | 12 | 2584 | |||
XM_017018439.1 | XP_016873928.1 | 13 | 2710 | |||
XM_017018440.1 | XP_016873929.1 | 14 | 2946 | |||
GenBank mRNA | AK027333.1 | - | 1 | 1062 | ||
AK055450.1 | - | 3 | 2287 | |||
AK090923.1 | - | 16 | 2953 | |||
AK127312.1 | - | 15 | 3859 | |||
AL832541.1 | - | 15 | 3119 | |||
BC039897.1 | - | 14 | 2784 | |||
BC046435.1 | - | 5 | 1600 | |||
BC064807.1 | - | 15 | 2885 | 5726473 |