Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267623.1 | NP_001254552.1 | 6 | 967 | ||
NM_001324236.1 | NP_001311165.1 | 7 | 1093 | |||
NM_001324237.1 | NP_001311166.1 | 8 | 998 | |||
NM_001324238.1 | NP_001311167.1 | 7 | 1001 | |||
NM_033453.3 | NP_258412.1 | 8 | 1090 | |||
NM_181493.3 | NP_852470.1 | 8 | 1039 | |||
NR_052000.1 | 7 | 933 | ||||
NR_052001.2 | 7 | 1011 | ||||
NR_052002.1 | 7 | 1025 | ||||
XM_006723564.3 | XP_006723627.1 | 9 | 1420 | |||
XM_006723565.3 | XP_006723628.1 | 7 | 1297 | |||
GenBank mRNA | AB062127.1 | - | 8 | 959 | ||
AF026816.2 | - | 8 | 949 | |||
AF063607.1 | - | 8 | 974 | |||
AF219116.1 | - | 8 | 1002 | |||
AF334680.1 | - | 7 | 882 | |||
BC010138.2 | - | 8 | 934 | 4124624 |