Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKX2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145285.2 | NP_660328.2 | 1 | 365 | ||
XM_011539370.1 | XP_011537672.1 | 1 | 356 | |||
GenBank mRNA | AK300856.1 | - | 1 | 317 | ||
AK313485.1 | - | 1 | 337 | |||
BC025788.1 | - | 1 | 345 | 5229146 | ||
BI764885.1 | - | 1 | 364 |