Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHE22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152414.4 | NP_689627.1 | 1 | 2061 | ||
GenBank mRNA | AK091333.1 | - | 1 | 1135 |