Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEIS2 SIRNA Assays ›
Gene Symbol:
MEIS2
Gene Name:
Meis homeobox 2
Gene Aliases:
HsT18361, MRG1
Chromosome Location:
Chr.15: 36891021 - 37101299 on Build GRCh38
UniGene ID:
Hs.510989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115086
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEIS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001220482.1
NP_001207411.1
12
1597
NM_002399.3
NP_002390.1
11
1452
NM_170674.4
NP_733774.1
11
1730
NM_170675.4
NP_733775.1
11
1751
NM_170676.4
NP_733776.1
11
1730
NM_170677.4
NP_733777.1
11
1751
NM_172315.2
NP_758526.1
10
1227
NM_172316.2
NP_758527.1
11
1517
NR_051953.1
10
1653
XM_006720523.1
XP_006720586.1
11
1141
XM_006720524.1
XP_006720587.1
11
1141
XM_006720525.1
XP_006720588.1
11
1120
XM_006720526.2
XP_006720589.1
11
2137
XM_006720527.2
XP_006720590.1
10
2018
XM_006720529.2
XP_006720592.1
10
2018
XM_011521591.2
XP_011519893.1
9
1203
XM_017022205.1
XP_016877694.1
11
2116
GenBank mRNA
AF017418.1
-
4
248
AF178948.1
-
11
1332
AF179896.1
-
11
1311
AF179897.1
-
11
1332
AF179898.1
-
11
1311
AF179899.1
-
10
1234
AK001298.1
-
2
433
AK055936.1
-
10
951
AK056038.1
-
10
1168
AK056216.1
-
11
2150
AK056620.1
-
11
1505
AK226072.1
-
11
1754
AK294411.1
-
9
905
AK300247.1
-
11
1485
AK311546.1
-
10
889
AY349358.1
-
2
165
BC001516.2
-
11
1456
2964406
BC001844.2
-
11
1456
2964406
BC007202.1
-
11
1456
2964406
BC050431.1
-
12
1477
6066728
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
eye development
transcription from RNA polymerase II promoter
visual learning
response to mechanical stimulus
pancreas development
negative regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
response to growth factor
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
transcription corepressor activity
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline