Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC9 SIRNA Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
UniGene ID:
Hs.196054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
115114
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204144.2
NP_001191073.1
5
569
NM_001204145.2
NP_001191074.1
3
261
NM_001204146.2
NP_001191075.1
3
430
NM_001204147.2
NP_001191076.1
2
227
NM_001204148.2
NP_001191077.1
2
227
NM_001321868.1
NP_001308797.1
4
509
NM_001321869.1
NP_001308798.1
4
509
NM_001321870.1
NP_001308799.1
4
509
NM_001321871.1
NP_001308800.1
4
509
NM_001321872.1
NP_001308801.1
4
509
NM_001321873.1
NP_001308802.1
4
509
NM_001321874.1
NP_001308803.1
3
430
NM_001321875.1
NP_001308804.1
3
430
NM_001321876.1
NP_001308805.1
3
430
NM_001321877.1
NP_001308806.1
3
430
NM_001321878.1
NP_001308807.1
3
430
NM_001321879.1
NP_001308808.1
3
430
NM_001321884.1
NP_001308813.1
3
430
NM_001321885.1
NP_001308814.1
3
430
NM_001321886.1
NP_001308815.1
4
489
NM_001321887.1
NP_001308816.1
4
320
NM_001321888.1
NP_001308817.1
3
261
NM_001321889.1
NP_001308818.1
4
489
NM_001321890.1
NP_001308819.1
3
430
NM_001321891.1
NP_001308820.1
3
430
NM_001321893.1
NP_001308822.1
4
327
NM_001321894.1
NP_001308823.1
4
327
NM_001321895.1
NP_001308824.1
3
261
NM_001321896.1
NP_001308825.1
4
599
NM_001321897.1
NP_001308826.1
4
327
NM_001321898.1
NP_001308827.1
3
261
NM_001321899.1
NP_001308828.1
4
327
NM_001321900.1
NP_001308829.1
3
430
NM_001321901.1
NP_001308830.1
3
261
NM_001321902.1
NP_001308831.1
3
261
NM_014707.3
NP_055522.1
3
261
NM_058176.2
NP_478056.1
2
116
NM_178423.2
NP_848510.1
3
261
NM_178425.3
NP_848512.1
2
116
XM_011515629.2
XP_011513931.1
3
210
XM_011515631.2
XP_011513933.1
4
509
XM_011515634.2
XP_011513936.1
3
210
XM_011515635.2
XP_011513937.1
4
1061
XM_011515636.2
XP_011513938.1
3
791
XM_011515637.2
XP_011513939.1
3
215
XM_011515638.2
XP_011513940.1
3
791
XM_011515639.2
XP_011513941.1
3
988
XM_011515641.2
XP_011513943.1
4
1058
XM_011515642.2
XP_011513944.1
2
1787
XM_011515644.2
XP_011513946.1
4
1048
XM_011515645.2
XP_011513947.1
2
1787
XM_011515646.2
XP_011513948.1
2
1787
XM_011515648.2
XP_011513950.1
3
791
XM_011515649.2
XP_011513951.1
3
995
XM_011515651.1
XP_011513953.1
3
191
XM_017012826.1
XP_016868315.1
3
791
XM_017012827.1
XP_016868316.1
4
850
XM_017012828.1
XP_016868317.1
3
990
XM_017012829.1
XP_016868318.1
3
191
XM_017012830.1
XP_016868319.1
3
191
XM_017012831.1
XP_016868320.1
3
191
XM_017012832.1
XP_016868321.1
3
191
GenBank mRNA
AB018287.1
-
3
225
AJ459808.1
-
3
225
AK097178.1
-
3
210
AK296214.1
-
3
162
AK297404.1
-
3
215
AK304298.1
-
5
562
AK304343.1
-
2
147
AK304410.1
-
2
190
AK316109.1
-
3
381
AY032737.1
-
2
75
AY032738.1
-
2
75
AY197371.1
-
2
75
AY429540.1
-
3
315
BC111735.1
-
2
74
BC150328.1
-
3
236
8860495
BC152405.1
-
3
225
8862508
CR749550.1
-
2
183
DC333353.1
-
4
509
DC333611.1
-
3
430
DC369193.1
-
2
223
DC369899.1
-
2
227
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001200045, XM_008764615, XM_008764610, XM_008764613, XM_008764616, and others.
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline