Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOSPHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143804.1 | NP_001137276.1 | 3 | 603 | ||
NM_178500.3 | NP_848595.1 | 3 | 476 | |||
XM_017024271.1 | XP_016879760.1 | 3 | 396 | |||
GenBank mRNA | AJ457189.1 | - | 3 | 500 | ||
BC117187.1 | - | 2 | 143 | 40125738 | ||
BC143884.1 | - | 2 | 270 |