Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSMEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134468.1 | NP_001127940.1 | 4 | 1433 | ||
NM_182597.2 | NP_872403.1 | 4 | 1639 | |||
XM_011516074.1 | XP_011514376.1 | 4 | 2142 | |||
XM_011516075.2 | XP_011514377.1 | 4 | 3829 | |||
XM_011516076.2 | XP_011514378.1 | 4 | 3465 | |||
XM_017012028.1 | XP_016867517.1 | 3 | 3347 | |||
GenBank mRNA | AK096894.1 | - | 4 | 1655 |