Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197975.2 | NP_932079.1 | 8 | 2061 | ||
GenBank mRNA | AB020625.1 | - | 8 | 1949 | ||
AK025267.1 | - | 8 | 2039 | |||
AY358385.1 | - | 8 | 2010 |