Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000168.5 | NP_000159.3 | 15 | 4866 | ||
GenBank mRNA | AK299299.1 | - | 14 | 4752 | ||
M57609.1 | - | 14 | 4830 |