Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000635.3 | NP_000626.2 | 18 | 2608 | ||
NM_134433.2 | NP_602309.1 | 17 | 2533 | |||
XM_011528171.2 | XP_011526473.1 | 19 | 2574 | |||
XM_011528172.1 | XP_011526474.1 | 18 | 2435 | |||
XM_017027107.1 | XP_016882596.1 | 17 | 2458 | |||
XM_017027108.1 | XP_016882597.1 | 16 | 2383 | |||
GenBank mRNA | AK024288.1 | - | 15 | 2147 | ||
AK093977.1 | - | 10 | 3163 | |||
AK291196.1 | - | 17 | 2412 | |||
AK302661.1 | - | 12 | 1593 | |||
AK303321.1 | - | 15 | 1917 | |||
BC028579.1 | - | 18 | 2520 | 4829582 | ||
BC071571.1 | - | 18 | 2489 | 5276369 | ||
BX648130.1 | - | 18 | 2443 | |||
X76091.1 | - | 18 | 2498 |