Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001455.3 | NP_001446.1 | 2 | 990 | ||
NM_201559.2 | NP_963853.1 | 3 | 963 | |||
XM_005266867.4 | XP_005266924.1 | 3 | 211 | |||
XM_005266868.3 | XP_005266925.1 | 2 | 235 | |||
XM_011535626.1 | XP_011533928.1 | 2 | 161 | |||
XM_011535628.2 | XP_011533930.1 | 2 | 4519 | |||
XM_011535629.2 | XP_011533931.1 | 2 | 184 | |||
XM_017010585.1 | XP_016866074.1 | 3 | 1896 | |||
XM_017010586.1 | XP_016866075.1 | 2 | 1779 | |||
GenBank mRNA | AF032886.1 | - | 2 | 1571 | ||
AJ001589.1 | - | 2 | 723 | |||
AK092357.1 | - | 3 | 351 | |||
AK122861.1 | - | 3 | 546 | |||
AK301304.1 | - | 2 | 152 | |||
AK303933.1 | - | 2 | 136 | |||
AK311587.1 | - | 2 | 250 | |||
BC020227.1 | - | 3 | 951 | 4111403 | ||
BC021224.2 | - | 3 | 951 | 4137370 | ||
BC068552.1 | - | 2 | 990 | 30347471 | ||
CR749261.1 | - | 2 | 220 |
Gene Symbol: FOXO3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026718.1 | 1 | 902 |