Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001537.3 | NP_001528.1 | 2 | 268 | ||
GenBank mRNA | AF068754.1 | - | 2 | 148 | ||
AK172732.1 | - | 2 | 133 | |||
BC007515.2 | - | 2 | 140 | 3010737 | ||
BX366956.2 | - | 2 | 200 | |||
BX537440.1 | - | 2 | 155 |