Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001537.3 | NP_001528.1 | 3, 4 | 400 | ||
GenBank mRNA | AF068754.1 | - | 3, 4 | 280 | ||
AK172732.1 | - | 3, 4 | 265 | |||
AY320409.1 | - | 3, 4 | 285 | |||
BC007515.2 | - | 3, 4 | 272 | 3010737 | ||
BX366956.2 | - | 3, 4 | 332 | |||
BX537440.1 | - | 3, 4 | 287 | |||
BX648972.1 | - | 3, 4 | 284 |