Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 4 | 365 | ||
NM_001321081.1 | NP_001308010.1 | 5 | 429 | |||
NM_001991.4 | NP_001982.2 | 5 | 456 | |||
XM_011524517.2 | XP_011522819.1 | 4 | 304 | |||
XM_017024350.1 | XP_016879839.1 | 4 | 283 | |||
XM_017024352.1 | XP_016879841.1 | 5 | 400 | |||
GenBank mRNA | AB002386.1 | - | 4 | 368 | ||
AB004818.1 | - | 4 | 374 | |||
AK295626.1 | - | 4 | 318 | |||
AK304835.1 | - | 4 | 269 | |||
AK309121.1 | - | 5 | 397 | |||
AL833159.1 | - | 5 | 376 | |||
BC015882.1 | - | 5 | 390 | 3908776 | ||
BT009782.1 | - | 3 | 268 | |||
DC333017.1 | - | 5 | 413 | |||
DC423942.1 | - | 5 | 456 | |||
U50315.1 | - | 4 | 374 |