Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 20 | 3566 | ||
NM_001321081.1 | NP_001308010.1 | 21 | 3630 | |||
NM_001321082.1 | NP_001308011.1 | 20 | 3537 | |||
NM_001991.4 | NP_001982.2 | 21 | 3657 | |||
XM_005257145.2 | XP_005257202.1 | 19 | 3412 | |||
XM_011524517.2 | XP_011522819.1 | 20 | 3505 | |||
XM_017024350.1 | XP_016879839.1 | 20 | 3484 | |||
XM_017024351.1 | XP_016879840.1 | 19 | 3391 | |||
XM_017024352.1 | XP_016879841.1 | 18 | 3242 | |||
GenBank mRNA | AB002386.1 | - | 20 | 3569 | ||
AL833159.1 | - | 21 | 3577 | |||
BC015882.1 | - | 21 | 3592 | 3908776 |