Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169122.1 | NP_001162593.1 | 10 | 2221 | ||
NM_001169123.1 | NP_001162594.1 | 11 | 2290 | |||
NM_001169124.1 | NP_001162595.1 | 10 | 2215 | |||
NM_001169125.1 | NP_001162596.1 | 10 | 2203 | |||
NM_001170628.1 | NP_001164099.1 | 8 | 854 | |||
NM_002025.3 | NP_002016.2 | 11 | 2320 | |||
GenBank mRNA | AB102644.1 | - | 10 | 1724 | ||
AJ001550.1 | - | 10 | 1724 | |||
AK301927.1 | - | 8 | 854 | |||
BC132683.1 | - | 11 | 1862 | 40146705 | ||
BC143740.1 | - | 10 | 1775 | 9052254 | ||
BC143744.1 | - | 10 | 1787 | 9052258 | ||
L76569.1 | - | 10 | 2134 | |||
U48436.1 | - | 11 | 2320 |