Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042539.2 | NP_001036004.1 | 2 | 992 | ||
NM_001276275.1 | NP_001263204.1 | 3 | 807 | |||
NM_002383.3 | NP_002374.2 | 2 | 992 | |||
XM_006721047.3 | XP_006721110.1 | 3 | 779 | |||
XM_006721048.3 | XP_006721111.1 | 2 | 1152 | |||
GenBank mRNA | AB209061.1 | - | 1 | 123 | ||
AF489858.1 | - | 2 | 939 | |||
BC041629.1 | - | 1 | 142 | 4309842 | ||
D85131.1 | - | 3 | 993 | |||
EF059746.1 | - | 2 | 827 | |||
FJ532357.1 | - | 3 | 797 | |||
L01420.1 | - | 1 | 239 | |||
M93339.1 | - | 2 | 878 | |||
M94046.1 | - | 2 | 849 | |||
U33819.1 | - | 2 | 1007 |