Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278610.1 | NP_001265539.1 | 3, 4 | 474 | ||
NM_002466.3 | NP_002457.1 | 4, 5 | 546 | |||
GenBank mRNA | AK223482.1 | - | 4, 5 | 281 | ||
AK303249.1 | - | 3, 4 | 379 | |||
AK314791.1 | - | 4, 5 | 451 | |||
BC007585.1 | - | 4, 5 | 471 | 3162656 | ||
BC053555.1 | - | 4, 5 | 440 | 6140320 | ||
X13293.1 | - | 4, 5 | 408 |