Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127892.1 | NP_001121364.1 | 3 | 4723 | ||
NM_002968.2 | NP_002959.2 | 3 | 4613 | |||
XM_006721241.3 | XP_006721304.1 | 4 | 4688 | |||
XM_011523254.2 | XP_011521556.1 | 4 | 4714 | |||
GenBank mRNA | EU668343.1 | - | 3 | 4581 | ||
Y18265.1 | - | 3 | 4581 |