Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199149.1 | NP_001186078.1 | 16 | 2258 | ||
NM_001321121.1 | NP_001308050.1 | 16 | 2363 | |||
NM_001321122.1 | NP_001308051.1 | 19 | 2602 | |||
NM_002343.5 | NP_002334.2 | 16 | 2369 | |||
GenBank mRNA | AF332168.1 | - | 16 | 2076 | ||
AK093852.1 | - | 19 | 2605 | |||
AK290859.1 | - | 16 | 2114 | |||
AK292813.1 | - | 16 | 2114 | |||
AK298035.1 | - | 16 | 2258 | |||
AK303995.1 | - | 8 | 977 | |||
AK316330.1 | - | 16 | 2178 | |||
AY137470.1 | - | 16 | 2076 | |||
AY165046.1 | - | 16 | 2088 | |||
AY178998.2 | - | 16 | 2076 | |||
AY360320.1 | - | 16 | 2071 | |||
AY493417.1 | - | 16 | 2186 | |||
AY875691.1 | - | 16 | 2076 | |||
BC015822.1 | - | 16 | 2118 | 4251222 | ||
BC015823.1 | - | 16 | 2120 | 4294752 | ||
BC022347.1 | - | 16 | 2114 | 4251058 | ||
EU779935.1 | - | 16 | 2076 | |||
EU794669.1 | - | 16 | 2111 | |||
KF992208.1 | - | 16 | 2088 | |||
KT006756.1 | - | 16 | 2076 | |||
M18642.1 | - | 6 | 771 | |||
M83202.1 | - | 16 | 2083 | |||
M83205.1 | - | 16 | 2062 | |||
M93150.1 | - | 16 | 2093 | |||
U07643.1 | - | 16 | 2173 | |||
X52941.1 | - | 16 | 2070 | |||
X53961.1 | - | 16 | 2370 |